Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023184.3 | NP_075673.1 | 2 | 938 | ||
GenBank mRNA | AF317225.1 | - | 1 | 813 | ||
AK007479.1 | - | 1 | 140 | |||
AK009739.1 | - | 2 | 938 | |||
AK138824.1 | - | 2 | 977 | |||
AK142618.1 | - | 2 | 940 | |||
BC013486.1 | - | 1 | 858 | 4211067 |