Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023184.3 | NP_075673.1 | 3 | 1276 | ||
GenBank mRNA | AF317225.1 | - | 2 | 1151 | ||
AK007479.1 | - | 2 | 515 | |||
AK009739.1 | - | 3 | 1276 | |||
AK138824.1 | - | 3 | 1315 | |||
AK142618.1 | - | 3 | 1278 | |||
BC013486.1 | - | 2 | 1196 | 4211067 |