Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290807.1 | NP_001277736.1 | 5 | 448 | ||
NM_001290808.1 | NP_001277737.1 | 4 | 478 | |||
NM_025475.3 | NP_079751.1 | 5 | 548 | |||
GenBank mRNA | AK007106.1 | - | 5 | 450 | ||
AK010445.1 | - | 5 | 495 | |||
AK012086.1 | - | 5 | 547 | |||
AK012779.1 | - | 4 | 478 | |||
AK012960.1 | - | 5 | 495 | |||
AK132300.1 | - | 5 | 504 | |||
BC086677.1 | - | 5 | 486 | 30660268 |