Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175105.3 | NP_780314.1 | 1 | 689 | ||
GenBank mRNA | AB028148.1 | - | 1 | 443 | ||
AK006005.1 | - | 1 | 714 | |||
AK040045.1 | - | 1 | 458 | |||
AK075757.1 | - | 1 | 690 | |||
AK137326.1 | - | 1 | 733 | |||
BC037088.1 | - | 1 | 468 | |||
BC068103.1 | - | 2 | 338 | |||
BC115612.2 | - | 3 | 481 | |||
BC115613.2 | - | 3 | 481 |