Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eid3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025499.2 | NP_079775.2 | 1 | 1090 | ||
GenBank mRNA | AK006438.1 | - | 1 | 1059 | ||
AK132745.1 | - | 1 | 1055 | |||
AK161275.1 | - | 1 | 1090 | |||
AK161307.1 | - | 1 | 1090 | |||
BC099545.1 | - | 1 | 1057 | 30917551 |