Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002326.1 | - | 9 | 1068 | ||
AK009159.1 | - | 9 | 1079 | |||
AK084508.1 | - | 9 | 996 | |||
BC010827.1 | - | 9 | 1050 | 4219628 | ||
EF410126.1 | - | 479 |