Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ergic3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025516.4 | NP_079792.1 | 4 | 331 | ||
GenBank mRNA | AK009350.1 | - | 4 | 332 | ||
AK013942.1 | - | 4 | 331 | |||
AK050285.1 | - | 4 | 346 | |||
AK179598.1 | - | 1 | 40 | |||
BC043720.1 | - | 4 | 356 | 5375947 | ||
BC057130.1 | - | 4 | 350 | 6819420 |