Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301176.1 | NP_001288105.1 | 2 | 220 | ||
NM_025518.4 | NP_079794.1 | 2 | 220 | |||
GenBank mRNA | AK009391.1 | - | 2 | 192 | ||
AK077451.1 | - | 2 | 192 | |||
BC116934.1 | - | 2 | 138 | 40126253 | ||
BC116936.1 | - | 2 | 138 | 40126255 |