Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025522.5 | NP_079798.2 | 5 | 725 | ||
GenBank mRNA | AK009385.1 | - | 5 | 704 | ||
AK014100.1 | - | 5 | 680 | |||
BC016189.1 | - | 5 | 691 | 4482814 | ||
CF586466.1 | - | 3 | 614 |