Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025522.5 | NP_079798.2 | 2 | 269 | ||
GenBank mRNA | AK009385.1 | - | 2 | 248 | ||
AK014100.1 | - | 2 | 224 | |||
BC016189.1 | - | 2 | 235 | 4482814 | ||
CX223953.1 | - | 2 | 269 |