Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahnak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039959.2 | NP_001035048.1 | 4 | 560 | ||
NM_001286518.1 | NP_001273447.1 | 4 | 560 | |||
NM_009643.2 | NP_033773.1 | 4 | 560 | |||
GenBank mRNA | AK009866.1 | - | 4 | 583 | ||
AK045184.1 | - | 4 | 561 | |||
AK134304.1 | - | 4 | 1572 | |||
BC028439.1 | - | 3 | 425 | |||
CB321491.1 | - | 4 | 543 |