Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtfdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025542.2 | NP_079818.1 | 1, 2 | 157 | ||
GenBank mRNA | AK009901.1 | - | 1, 2 | 153 | ||
AK010321.1 | - | 1, 2 | 158 | |||
AK028792.1 | - | 1, 2 | 146 | |||
AK053258.1 | - | 1, 2 | 170 | |||
AK077823.1 | - | 1, 2 | 158 | |||
AK153121.1 | - | 1, 2 | 111 | |||
AK167287.1 | - | 1, 2 | 147 | |||
AK169588.1 | - | 1, 2 | 134 | |||
AK170141.1 | - | 1, 2 | 134 | |||
AK219307.1 | - | 1, 2 | 135 | |||
BC004803.1 | - | 1, 2 | 145 | 3589578 | ||
BC049072.1 | - | 1, 2 | 150 | 5701585 | ||
BC096692.1 | - | 1, 2 | 106 | 5255425 | ||
BC116442.1 | - | 1, 2 | 38 | |||
BC116443.1 | - | 1, 2 | 38 | |||
BC127166.1 | - | 1, 2 | 38 |