Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsl1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025546.2 | NP_079822.1 | 3 | 316 | ||
GenBank mRNA | AK012937.1 | - | 3 | 309 | ||
AK075933.1 | - | 3 | 317 | |||
AK136025.1 | - | 3 | 283 | |||
AK145699.1 | - | 3 | 330 | |||
AK162241.1 | - | 3 | 290 | |||
AK165572.1 | - | 3 | 329 | |||
AK166479.1 | - | 3 | 292 | |||
AK166579.1 | - | 3 | 312 | |||
AK168469.1 | - | 3 | 312 | |||
AK168717.1 | - | 3 | 305 | |||
AK171805.1 | - | 3 | 304 | |||
BC034355.1 | - | 3 | 282 | 2651674 | ||
BC069934.1 | - | 3 | 290 | |||
DQ445383.1 | - | 3 | 324 |