Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025553.4 | NP_079829.1 | 2 | 246 | ||
GenBank mRNA | AB049639.1 | - | 2 | 224 | ||
AK002797.1 | - | 2 | 201 | |||
AK010329.1 | - | 2 | 238 | |||
AK167247.1 | - | 2 | 214 | |||
BC046750.1 | - | 2 | 199 | 5692108 |