Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025579.3 | NP_079855.2 | 3 | 400 | ||
GenBank mRNA | AK007811.1 | - | 3 | 363 | ||
AK013133.1 | - | 3 | 363 | |||
AK132927.1 | - | 3 | 401 | |||
AK139748.1 | - | 3 | 314 | |||
BC019668.1 | - | 3 | 345 | 3493047 |