Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025609.2 | NP_079885.2 | 3 | 209 | ||
GenBank mRNA | AB088136.1 | - | 3 | 209 | ||
BC027054.1 | - | 3 | 201 | |||
BC041110.1 | - | 3 | 204 | |||
BC054369.1 | - | 3 | 209 | 5356886 | ||
BC114637.1 | - | 3 | 230 |