Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmigd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025655.2 | NP_079931.1 | 1 | 64 | ||
GenBank mRNA | AK008028.1 | - | 2 | 125 | ||
AK009118.1 | - | 1 | 64 | |||
BC022683.1 | - | 1 | 31 | |||
BC061082.1 | - | 1 | 54 | 30311543 |