Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110809.2 | NP_001104279.1 | 4, 5 | 379 | ||
NM_001286857.1 | NP_001273786.1 | 4, 5 | 993 | |||
NM_025663.4 | NP_079939.3 | 5, 6 | 514 | |||
GenBank mRNA | AK011617.1 | - | 4, 5 | 378 | ||
AK017310.1 | - | 4, 5 | 329 | |||
AK018306.1 | - | 4, 5 | 353 | |||
AK078828.1 | - | 4, 5 | 377 | |||
AK169111.1 | - | 4, 5 | 368 | |||
BC019490.1 | - | 5, 6 | 496 | 4039649 | ||
BC080676.1 | - | 4, 5 | 971 | |||
BI658293.1 | - | 5, 6 | 514 |