Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174874.3 | NP_777363.1 | 8, 9 | 743 | ||
GenBank mRNA | AJ504653.1 | - | 7, 8 | 735 | ||
AK028712.1 | - | 8, 9 | 754 | |||
AK031570.1 | - | 8, 9 | 744 | |||
AK075796.1 | - | 8, 9 | 754 | |||
AK088811.1 | - | 8, 9 | 816 | |||
AK129242.1 | - | 6, 7 | 3315 | |||
AK144239.1 | - | 8, 9 | 749 | |||
BC027184.1 | - | 8, 9 | 769 | 4237950 |