Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174874.3 | NP_777363.1 | 6, 7 | 472 | ||
GenBank mRNA | AJ504653.1 | - | 5, 6 | 464 | ||
AK010949.1 | - | 6, 7 | 600 | |||
AK028712.1 | - | 6, 7 | 483 | |||
AK031570.1 | - | 6, 7 | 473 | |||
AK075796.1 | - | 6, 7 | 483 | |||
AK088811.1 | - | 6, 7 | 545 | |||
AK144239.1 | - | 6, 7 | 478 | |||
AK188147.1 | - | 4, 5 | 321 | |||
AK204065.1 | - | 2, 3 | 160 | |||
BC027184.1 | - | 6, 7 | 498 | 4237950 |