Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025666.2 | NP_079942.1 | 8 | 991 | ||
GenBank mRNA | AK088208.1 | - | 8 | 993 | ||
AK146308.1 | - | 8 | 993 | |||
AK149989.1 | - | 8 | 946 | |||
AK155611.1 | - | 8 | 862 | |||
AK201594.1 | - | 2 | 354 | |||
AK203922.1 | - | 3 | 354 | |||
BC051678.1 | - | 5 | 519 | |||
BC058535.1 | - | 7 | 675 | |||
BC079531.1 | - | 7 | 688 | |||
BC094323.1 | - | 8 | 998 | |||
BC094423.1 | - | 8 | 949 | 6812389 |