Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hiatl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083901.1 | NP_001077370.1 | 7 | 801 | ||
NM_133680.3 | NP_598441.3 | 6 | 898 | |||
GenBank mRNA | AK146596.1 | - | 7 | 794 | ||
BC033469.1 | - | 6 | 860 | 4951631 | ||
CN709077.1 | - | 3 | 227 |