Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285429.1 | NP_001272358.1 | 4 | 342 | ||
NM_001285430.1 | NP_001272359.1 | 4 | 342 | |||
NM_001285431.1 | NP_001272360.1 | 3 | 270 | |||
NM_001285432.1 | NP_001272361.1 | 3 | 273 | |||
NM_001285433.1 | NP_001272362.1 | 3 | 270 | |||
NM_001285434.1 | NP_001272363.1 | 4 | 327 | |||
NM_023240.3 | NP_075729.2 | 4 | 345 | |||
NM_029663.2 | NP_083939.1 | 6 | 1490 | |||
GenBank mRNA | AF304351.1 | - | 4 | 308 | ||
AK010347.1 | - | 5 | 320 | |||
AK017616.1 | - | 3 | 225 | |||
AK017796.1 | - | 4 | 308 | |||
AK044934.1 | - | 4 | 297 | |||
BC013059.1 | - | 4 | 275 | |||
BC079855.1 | - | 6 | 1436 | 6403006 | ||
BC082536.1 | - | 6 | 1421 | |||
BQ177630.1 | - | 4 | 312 | |||
DV062370.1 | - | 3 | 277 |