Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285429.1 | NP_001272358.1 | 2 | 168 | ||
NM_001285430.1 | NP_001272359.1 | 2 | 168 | |||
NM_001285431.1 | NP_001272360.1 | 2 | 168 | |||
NM_001285432.1 | NP_001272361.1 | 2 | 171 | |||
NM_001285433.1 | NP_001272362.1 | 2 | 168 | |||
NM_001285434.1 | NP_001272363.1 | 2 | 168 | |||
NM_023240.3 | NP_075729.2 | 2 | 171 | |||
NM_029663.2 | NP_083939.1 | 4 | 1316 | |||
GenBank mRNA | AF304351.1 | - | 2 | 134 | ||
AK010347.1 | - | 3 | 146 | |||
AK017616.1 | - | 2 | 123 | |||
AK017796.1 | - | 2 | 134 | |||
AK044934.1 | - | 2 | 123 | |||
AK054335.1 | - | 2 | 125 | |||
BC013059.1 | - | 2 | 116 | |||
BC079855.1 | - | 4 | 1262 | 6403006 | ||
BC082536.1 | - | 4 | 1249 | |||
BQ177630.1 | - | 2 | 138 | |||
BY035199.1 | - | 2 | 173 | |||
BY104511.1 | - | 2 | 159 | |||
BY154960.1 | - | 2 | 170 | |||
DV062370.1 | - | 2 | 175 |