Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025691.1 | NP_079967.1 | 1 | 72 | ||
GenBank mRNA | AK133784.1 | - | 1 | 73 | ||
AK138875.1 | - | 1 | 60 | |||
BC003434.1 | - | 1 | 60 | |||
BC019196.1 | - | 1 | 65 |