Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023243.5 | NP_075732.1 | 5 | 612 | ||
GenBank mRNA | AF287135.1 | - | 5 | 665 | ||
AK007910.1 | - | 2 | 77 | |||
AK078059.1 | - | 5 | 613 | |||
AK137849.1 | - | 5 | 641 | |||
BC038861.1 | - | 5 | 591 | 1446473 |