Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025700.2 | NP_079976.1 | 12 | 1626 | ||
GenBank mRNA | AK004286.1 | - | 3 | 307 | ||
AK014332.1 | - | 12 | 1551 | |||
BC030869.1 | - | 12 | 1588 | |||
BC052762.1 | - | 12 | 1626 | 6510389 |