Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcbld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025705.3 | NP_079981.2 | 8, 9 | 941 | ||
GenBank mRNA | AK014521.1 | - | 8, 9 | 753 | ||
AK016485.1 | - | 8, 9 | 941 | |||
BC026771.1 | - | 7, 8 | 729 | 4014778 |