Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gid4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025757.4 | NP_080033.3 | 5 | 989 | ||
GenBank mRNA | AJ404329.1 | - | 5 | 477 | ||
AK012042.1 | - | 5 | 817 | |||
AK017120.1 | - | 989 | ||||
AK039435.1 | - | 5 | 873 | |||
AK077759.1 | - | 5 | 556 | |||
AK142753.1 | - | 5 | 791 | |||
AK164644.1 | - | 5 | 856 | |||
BC044901.1 | - | 5 | 691 | 3660276 | ||
BC046821.1 | - | 5 | 803 | 6400140 | ||
BC052418.1 | - | 5 | 831 | 6401069 |