Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam187a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025766.2 | NP_080042.1 | 1 | 311 | ||
GenBank mRNA | AK016939.1 | - | 1 | 312 | ||
AK017119.1 | - | 1 | 312 | |||
BC116769.1 | - | 1 | 157 | 40126088 | ||
BC116771.1 | - | 1 | 157 | 40126090 |