Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025768.2 | NP_080044.2 | 7 | 914 | ||
GenBank mRNA | AF329833.1 | - | 5 | 702 | ||
AK007905.1 | - | 7 | 919 | |||
AK017197.1 | - | 7 | 878 | |||
AK017251.1 | - | 8 | 876 | |||
AK082863.1 | - | 7 | 908 | |||
AK149322.1 | - | 911 | ||||
BC029096.1 | - | 7 | 920 | 3500563 |