Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntnap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004357.2 | NP_001004357.2 | 23 | 3983 | ||
NM_025771.3 | NP_080047.1 | 3 | 430 | |||
GenBank mRNA | AK013139.1 | - | 3 | 398 | ||
AK017341.1 | - | 3 | 425 | |||
AK129232.1 | - | 23 | 3956 | |||
BC029826.1 | - | 3 | 379 | |||
BC064467.1 | - | 18 | 2874 | |||
BC103799.1 | - | 3 | 337 | |||
BC132074.1 | - | 2 | 242 | |||
BC132076.1 | - | 2 | 242 |