Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173547.2 | NP_001167018.1 | 7, 8 | 839 | ||
NM_181516.6 | NP_852657.1 | 7, 8 | 836 | |||
GenBank mRNA | AK075615.1 | - | 7, 8 | 793 | ||
AK076481.1 | - | 7, 8 | 804 | |||
AK178858.1 | - | 4, 5 | 254 | |||
AK183640.1 | - | 2, 3 | 368 | |||
AK188231.1 | - | 5, 6 | 294 | |||
AY231459.1 | - | 7, 8 | 555 | |||
AY258041.1 | - | 7, 8 | 555 | |||
BC015305.1 | - | 7, 8 | 801 | 4221041 |