Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173547.2 | NP_001167018.1 | 5, 6 | 732 | ||
NM_001242615.2 | NP_001229544.1 | 5, 6 | 732 | |||
NM_001242616.2 | NP_001229545.1 | 5, 6 | 732 | |||
NM_181516.6 | NP_852657.1 | 5, 6 | 732 | |||
GenBank mRNA | AK075615.1 | - | 5, 6 | 689 | ||
AK076481.1 | - | 5, 6 | 700 | |||
AK160327.1 | - | 5, 6 | 701 | |||
AK170662.1 | - | 5, 6 | 701 | |||
AK178858.1 | - | 2, 3 | 150 | |||
AK183640.1 | - | 1, 2 | 32 | |||
AK188231.1 | - | 3, 4 | 190 | |||
AY231459.1 | - | 5, 6 | 451 | |||
AY231460.1 | - | 5, 6 | 451 | |||
AY258040.1 | - | 5, 6 | 451 | |||
AY258041.1 | - | 5, 6 | 451 | |||
BC015305.1 | - | 5, 6 | 697 | 4221041 | ||
EF524069.1 | - | 4, 5 | 331 |