Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025793.3 | NP_080069.2 | 3 | 321 | ||
GenBank mRNA | AK002352.1 | - | 3 | 282 | ||
AK013170.1 | - | 3 | 271 | |||
AK146101.1 | - | 3 | 322 | |||
AK151975.1 | - | 3 | 283 | |||
AK168603.1 | - | 3 | 274 | |||
AK170602.1 | - | 3 | 268 | |||
AK182151.1 | - | 169 | ||||
AK188918.1 | - | 2 | 120 | |||
AK189483.1 | - | 3 | 121 | |||
AK215641.1 | - | 3 | 120 | |||
BC004595.1 | - | 3 | 265 | 3584839 |