Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025799.4 | NP_080075.2 | 1 | 283 | ||
GenBank mRNA | AK002675.1 | - | 1 | 225 | ||
AK008653.1 | - | 1 | 228 | |||
AK017443.1 | - | 1 | 254 | |||
AK030320.1 | - | 1 | 285 | |||
AK035607.1 | - | 1 | 164 | |||
AK044790.1 | - | 1 | 140 | |||
AK146295.1 | - | 1 | 212 | |||
AK151204.1 | - | 1 | 144 | |||
AK159288.1 | - | 1 | 229 | |||
AK163042.1 | - | 1 | 227 | |||
AK179693.1 | - | 1 | 28 | |||
BC004039.1 | - | 1 | 234 | 3496187 | ||
BC018181.1 | - | 1 | 243 | 3983067 | ||
BC094449.1 | - | 1 | 198 | 4222249 |