Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tanc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290659.1 | NP_001277588.1 | 26 | 4430 | ||
NM_198294.3 | NP_938036.2 | 27 | 4606 | |||
GenBank mRNA | AB093299.1 | - | 16 | 2699 | ||
AK004560.1 | - | 2 | 559 | |||
BC047437.1 | - | 6 | 796 | |||
BC079914.1 | - | 26 | 4409 | 30668628 | ||
BC141367.1 | - | 26 | 4420 | 9055980 |