Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tanc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290659.1 | NP_001277588.1 | 8 | 1134 | ||
NM_198294.3 | NP_938036.2 | 9 | 1289 | |||
GenBank mRNA | AK030022.1 | - | 6 | 893 | ||
BC079914.1 | - | 8 | 1092 | 30668628 | ||
BC141367.1 | - | 8 | 1124 | 9055980 |