Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acadsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025826.4 | NP_080102.1 | 10 | 1287 | ||
GenBank mRNA | AK004889.1 | - | 10 | 1188 | ||
AK076535.1 | - | 10 | 1189 | |||
AK152325.1 | - | 10 | 1176 | |||
AK163322.1 | - | 6 | 768 | |||
BC011235.1 | - | 10 | 1169 | |||
BC035231.1 | - | 10 | 1153 | |||
BC044826.1 | - | 10 | 1220 | |||
BC054428.1 | - | 10 | 1229 | 5039627 |