Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lman2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025828.3 | NP_080104.2 | 5 | 843 | ||
GenBank mRNA | AK004952.1 | - | 5 | 674 | ||
AK083442.1 | - | 5 | 675 | |||
AK129028.1 | - | 5 | 670 | |||
AK150383.1 | - | 5 | 672 | |||
AK166953.1 | - | 5 | 680 | |||
AK169401.1 | - | 5 | 671 | |||
BC049221.1 | - | 5 | 667 | 2599070 | ||
BC055327.1 | - | 5 | 843 | 5702182 |