Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025836.3 | NP_080112.1 | 8 | 1356 | ||
GenBank mRNA | AK003971.1 | - | 1 | 209 | ||
AK004970.1 | - | 8 | 1357 | |||
AK009148.1 | - | 4 | 902 | |||
AK012517.1 | - | 8 | 1357 | |||
AK049537.1 | - | 8 | 1358 | |||
AK050140.1 | - | 8 | 1358 | |||
AK132313.1 | - | 1357 | ||||
AK167214.1 | - | 8 | 1355 | |||
AK200250.1 | - | 1 | 50 | |||
AK201044.1 | - | 1 | 49 | |||
AK210643.1 | - | 1 | 49 | |||
AK213759.1 | - | 1 | 49 | |||
AK214545.1 | - | 1 | 49 | |||
BC011116.1 | - | 8 | 1352 | 4023354 | ||
DQ257436.1 | - | 8 | 979 |