Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025836.3 | NP_080112.1 | 7 | 993 | ||
GenBank mRNA | AK004970.1 | - | 7 | 994 | ||
AK009148.1 | - | 3 | 540 | |||
AK012517.1 | - | 7 | 994 | |||
AK049537.1 | - | 7 | 994 | |||
AK050140.1 | - | 7 | 995 | |||
AK132313.1 | - | 994 | ||||
AK167214.1 | - | 7 | 992 | |||
BC011116.1 | - | 7 | 989 | 4023354 | ||
DQ257436.1 | - | 7 | 748 |