Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chordc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025844.2 | NP_080120.1 | 6 | 579 | ||
GenBank mRNA | AK003259.1 | - | 6 | 580 | ||
AK132696.1 | - | 559 | ||||
BC018374.1 | - | 6 | 579 | 4236535 |