Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024184.2 | NP_077146.1 | 3 | 453 | ||
GenBank mRNA | AK005646.1 | - | 3 | 486 | ||
AK019594.1 | - | 3 | 235 | |||
AK077005.1 | - | 3 | 479 | |||
AK077718.1 | - | 3 | 434 | |||
AK134906.1 | - | 3 | 641 | |||
AK150818.1 | - | 3 | 434 | |||
AK151951.1 | - | 3 | 434 | |||
AK165322.1 | - | 3 | 432 | |||
BC003428.1 | - | 3 | 453 | 2645378 |