Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025850.2 | NP_080126.1 | 10 | 1014 | ||
GenBank mRNA | AK005694.1 | - | 10 | 1024 | ||
AK017071.1 | - | 10 | 1038 | |||
AY819025.1 | - | 10 | 1044 | |||
BC046278.1 | - | 10 | 1014 | 6310911 |