Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700010I14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025851.3 | NP_080127.2 | 8 | 1353 | ||
GenBank mRNA | AK005830.1 | - | 8 | 1333 | ||
AK015220.1 | - | 10 | 1532 | |||
AK161278.1 | - | 8 | 1330 | |||
BC050753.1 | - | 8 | 1325 | |||
BC061078.1 | - | 8 | 1303 | 6770439 |