Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025894.2 | NP_080170.1 | 8 | 905 | ||
GenBank mRNA | AK005114.1 | - | 8 | 939 | ||
AK007619.1 | - | 8 | 950 | |||
AK007680.1 | - | 8 | 879 | |||
AK009969.1 | - | 8 | 908 | |||
AK017843.1 | - | 8 | 922 | |||
AK150910.1 | - | 8 | 928 | |||
AK151242.1 | - | 8 | 906 | |||
AK152041.1 | - | 8 | 928 | |||
AK152240.1 | - | 8 | 937 | |||
AK160283.1 | - | 8 | 927 | |||
AK162779.1 | - | 906 | ||||
AK167236.1 | - | 8 | 913 | |||
BC004694.1 | - | 8 | 923 | 3498952 |