Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025894.2 | NP_080170.1 | 10 | 1159 | ||
GenBank mRNA | AK005114.1 | - | 10 | 1193 | ||
AK007619.1 | - | 10 | 1204 | |||
AK007680.1 | - | 10 | 1133 | |||
AK008646.1 | - | 10 | 1158 | |||
AK009969.1 | - | 10 | 1162 | |||
AK017843.1 | - | 10 | 1176 | |||
AK150910.1 | - | 10 | 1182 | |||
AK151242.1 | - | 10 | 1160 | |||
AK152041.1 | - | 10 | 1182 | |||
AK152240.1 | - | 10 | 1191 | |||
AK160283.1 | - | 10 | 1182 | |||
AK162779.1 | - | 1160 | ||||
AK167236.1 | - | 10 | 1166 | |||
BC004694.1 | - | 10 | 1177 | 3498952 |