Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025894.2 | NP_080170.1 | 7, 8 | 839 | ||
GenBank mRNA | AK005114.1 | - | 7, 8 | 873 | ||
AK007619.1 | - | 7, 8 | 884 | |||
AK007680.1 | - | 7, 8 | 813 | |||
AK008646.1 | - | 7, 8 | 840 | |||
AK009969.1 | - | 7, 8 | 842 | |||
AK017843.1 | - | 7, 8 | 856 | |||
AK150910.1 | - | 7, 8 | 862 | |||
AK151242.1 | - | 7, 8 | 840 | |||
AK152041.1 | - | 7, 8 | 862 | |||
AK152240.1 | - | 7, 8 | 871 | |||
AK160283.1 | - | 7, 8 | 861 | |||
AK162779.1 | - | 840 | ||||
AK167236.1 | - | 7, 8 | 847 | |||
AK178211.1 | - | 1, 2 | 74 | |||
BC004694.1 | - | 7, 8 | 857 | 3498952 |