Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080554.2 | NP_542121.2 | 9 | 1274 | ||
GenBank mRNA | AK011026.1 | - | 9 | 1251 | ||
AK029067.1 | - | 7 | 2651 | |||
AK032992.1 | - | 9 | 1252 | |||
AK078419.1 | - | 9 | 1252 | |||
AK145587.1 | - | 9 | 1251 | |||
AK148284.1 | - | 1275 | ||||
AK208538.1 | - | 1 | 98 | |||
AK213478.1 | - | 1 | 98 | |||
AK220321.1 | - | 9 | 1215 | |||
BC010650.1 | - | 3 | 375 | |||
BC019112.1 | - | 9 | 1285 |